Learn More
Sino Biological Anti-Complement C1s Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10220MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement C1s (rh Complement C1s ; Catalog#10220-H08H; NP_001725.1; Met1-Asp688). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3317