Learn More
Sino Biological Recombinant Human Leptin Protein
Supplier:  Sino Biological 10221HNAE1
Encompass_Preferred
A DNA sequence encoding the mature form of human Leptin (NP_000221.1) (Val 22-Cys 167) was expressed, with an additional Met.
Catalog No. 50-161-4372