Learn More
Sino Biological Anti-Leptin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10221MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Leptin (rh Leptin; Catalog#10221-HNAE; NP_000221.1; Val22-Cys167). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3336