Learn More
Sino Biological Anti-Leptin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10221RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Leptin (rh Leptin; Catalog#10221-HNAE; NP_000221.1; Val 22-Cys 167). Leptin specific IgG was purified by human Leptin affinity chromatography.
Catalog No. 50-172-3348