Learn More
Sino Biological Anti-Lipocalin-2/NGAL Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10222MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LCN2 / NGAL (rh LCN2 / NGAL; Catalog#10222-H08H; NP_005555.2; Met1-Gly198). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.
Catalog No. 50-172-3360