Learn More
Sino Biological Anti-Napsin A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102221T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NAPSA/Napsin A, and purified by antigen affinity chromatography.
Catalog No. 50-172-3351