Learn More
Sino Biological Recombinant Human DcR3/TNFRSF6B Protein (hFc Tag)
Supplier:  Sino Biological 10224H02B100
Encompass_Preferred
A DNA sequence encoding the human DCR3 (O95407)(Met1-His300) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4378