Learn More
Sino Biological Anti-CYP3A4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102243T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CYP3A4 / Cytochrome P450 3A4, and purified by antigen affinity chromatography.
Catalog No. 50-172-3384