Learn More
Sino Biological Anti-NOS2/iNOS Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102279T02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NOS2, and purified by antigen affinity chromatography.
Catalog No. 50-172-3405