Learn More
Sino Biological Recombinant Human ACVR2B Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10229H02H100
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Thr 134) from the extracellular domain of human ACVR2B (NP_001097.2) was expressed with the fused human IgG1 Fc region at the C-terminus.
Catalog No. 50-161-4384