Learn More
Sino Biological Recombinant Human R-Cadherin/CDH4 Protein (His Tag)
Supplier:  Sino Biological 10230H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CAD4 (NP_001785.2) (Met 1-Ala 734) was expressed with a fused polyhistidine tag at the C-terminus.
Catalog No. 50-161-4388