Learn More
Sino Biological Anti-R-Cadherin/CDH4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10230RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CDH4 extracellular domain (rh CDH4; Catalog#10230-H08H; NP_001785.2; Met 1-Ala 734). CDH4 specific IgG was purified by human CDH4 affinity chromatography.
Catalog No. 50-172-3461