Learn More
Sino Biological Anti-R-Cadherin/CDH4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10230T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human R-Cadherin / CDH4 (rh R-Cadherin / CDH4; Catalog#10230-H08H; NP_001785.2; Met1-Ala734). R-Cadherin / CDH4 specific IgG was purified by Human R-Cadherin / CDH4 affinity chromatography.
Catalog No. 50-172-3464