Learn More
Sino Biological Recombinant Anti-LDLR/LDL Receptor Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10231R301100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human LDL Receptor / LDLR (rh LDL Receptor / LDLR; Catalog#10231-H08H; NP_000518.1; Met1-Arg788).
Catalog No. 50-172-3497