Learn More
Sino Biological Recombinant Anti-LDLR/LDL Receptor Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 10231R301P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human LDL Receptor / LDLR (rh LDL Receptor / LDLR; Catalog#10231-H08H; NP_000518.1; Met 1-Arg 788) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-3503