Learn More
Sino Biological Anti-F4/80/EMR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102310T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human EMR1, and purified by antigen affinity chromatography.
Catalog No. 50-172-3467