Learn More
Sino Biological Recombinant Human TARC/CCL17 Protein (His Tag)
Supplier:  Sino Biological 10233H08B50
Encompass_Preferred
A DNA sequence encoding the full length of human CCL17 (Q92583-1) (Met 1-Ser 94) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4393