Learn More
Sino Biological Anti-TARC/CCL17 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10233MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCL17 / TARC (rh CCL17 / TARC; Catalog#10233-HNAB; Q92583-1; Met 1-Ser 94). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3517