Learn More
Sino Biological Anti-TARC/CCL17 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10233RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CCL17 / TARC (rh CCL17; Catalog#10233-HNAB; Q92583-1; Met1-Ser94). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-3524