Learn More
Sino Biological Anti-TARC/CCL17 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10233RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CCL17 / TARC (rh CCL17 / TARC; Catalog#10233-HNAB; Q92583-1; Met 1-Ser 94). CCL17 / TARC specific IgG was purified by Human CCL17 / TARC affinity chromatography.
Catalog No. 50-172-3527