Learn More
Sino Biological Recombinant Human EphB4 Protein (hFc Tag)
Supplier:  Sino Biological 10235H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ala 539) of human EphB4 (NP_004435.3) precursor was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4396