Learn More
Sino Biological Recombinant Human EphB4 Protein (aa 563-987, His & GST Tag)
Supplier:  Sino Biological 10235H20B150
Encompass_Preferred
A DNA sequence encoding the human EPHB4 (P54760) (Leu563-Tyr987) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-4402