Learn More
Sino Biological Anti-EphB4 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10235MM04A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EphB4 (rh EphB4; Catalog#10235-H08H; NP_004435.3; Met 1-Ala 539) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-3549