Learn More
Sino Biological Anti-EphB4 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10235MM04F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EphB4 (rhEphB4; Catalog#10235-H08H; NP_004435.3; Met 1-Ala 539) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-3551