Learn More
Sino Biological Anti-EphB4 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10235MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EphB4 / Eph Receptor B4 (rh EphB4 / Eph Receptor B4; Catalog#10235-H08H; NP_004435.3; Met1-Ala539). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3554