Learn More
Sino Biological Recombinant Anti-EphB4 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10235R001A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB4 (rh EphB4; Catalog#10235-H08H; NP_004435.3; Met 1-Ala 539) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-3562