Learn More
Sino Biological Anti-RON/MSP receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102358T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse CD136/MST1R, and purified by antigen affinity chromatography.
Catalog No. 50-172-3542