Learn More
Sino Biological Recombinant Human BCAM Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10238H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human BCAM (NP_005572.2) (Met 1-Ala 547) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4407