Learn More
Sino Biological Recombinant Human CD40 Ligand/TNFSF5 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10239H08E100
Encompass_Preferred
A DNA sequence encoding the soluble form of human CD40L (NP_000065.1) (Glu 108-Leu 261) was expressed with an initial Met at the N-terminus and a polyhistide tag at the C-terminus.
Catalog No. 50-161-4414