Learn More
Sino Biological Anti-ANO1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102411T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Ano1, and purified by antigen affinity chromatography.
Catalog No. 50-172-3635