Learn More
Sino Biological Anti-Tubby Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102415T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human TUB/tubby bipartite transcription factor, and purified by antigen affinity chromatography.
Catalog No. 50-172-3638