Learn More
Sino Biological Anti-pro renin receptor/ATP6AP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102419T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human pro renin receptor/ATP6AP2, and purified by antigen affinity chromatography.
Catalog No. 50-172-3641