Learn More
Sino Biological Recombinant Human MD1 Protein (hFc Tag)
Supplier:  Sino Biological 10242H02H100
Encompass_Preferred
A DNA sequence encoding the human LY86 (O95711) (Met1-Ser162) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4421