Learn More
Sino Biological Recombinant Human methionyl aminopeptidase 2/METAP2 Protein (His Tag)
Supplier:  Sino Biological 10245H08B100
Encompass_Preferred
A DNA sequence encoding the human METAP2 (NP_006829.1) (Met1-Tyr478) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4425