Learn More
Sino Biological Anti-methionyl aminopeptidase 2/METAP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10245T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MAP2 / METAP2 (rh MAP2 / METAP2; Catalog#10245-H08B; NP_006829.1; Met1-Tyr478). MAP2 / METAP2 specific IgG was purified by Human MAP2 / METAP2 affinity chromatography.
Catalog No. 50-172-3668