Learn More
Sino Biological Anti-Midkine Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10247MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Midkine (rh Midkine; Catalog#10247-HNAB; P21741; Met1-Asp143). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3677