Learn More
Sino Biological Recombinant Human IL-11RA/IL-11R alpha Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10252H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IL11Rα (NP_004503.1) (Met 1-Val 363) was expressed, with a fused polyhistidine tag at the C-terminus.
Catalog No. 50-161-4436