Learn More
Sino Biological Anti-nkx6.1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102528T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NKX6-1, and purified by antigen affinity chromatography.
Catalog No. 50-172-3704