Learn More
Sino Biological Recombinant Anti-MMP-8 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10254R206100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human MMP-8 / MMP8 (rh MMP-8 / MMP8; Catalog#10254-H08H; NP_002415.1; Met1-Gly467).
Catalog No. 50-172-3720