Learn More
Sino Biological Anti-MMP-8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10254RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MMP8 (rh MMP8; Catalog#10254-H08H; NP_002415.1; Met 1-Gly 467). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-3722