Learn More
Sino Biological Recombinant Human CD64/FCGR1 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10256H27HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD64 (NP_000557.1) (Met 1-Thr 287) was fused with a C-terminal polyhistidine tagged AVI tag at the C-terminus The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-161-4450