Learn More
Sino Biological Anti-CD64/FCGR1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10256MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD64 (rh CD64; Catalog#10256-H08H; NP_000557.1; Met 1-Pro 288). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3740