Learn More
Sino Biological Recombinant Anti-CD64/FCGR1 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10256R401A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD64/Fc gamma RI (rh CD64/Fc gamma RI; Catalog#10256-H08H; NP_000557.1; Met1-Pro288) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-3746