Learn More
Sino Biological Recombinant Anti-CD64/FCGR1 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10256R407F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD64/Fc gamma RI (rh CD64/Fc gamma RI; Catalog#10256-H08H; NP_000557.1; Met1-Pro288) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-3753