Learn More
Sino Biological Recombinant Human ACVR2A Protein (hFc Tag)
Supplier:  Sino Biological 10257H02H200
Encompass_Preferred
A DNA sequence encoding the N-terminal segment (Met 1-Pro 134) from the extracellular domain of human ACVR2A (NP_001607.1) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4453