Learn More
Sino Biological Recombinant Human ACVR2A Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10257H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human ACVR2A (NP_001607.1) (Met 1-Pro 134) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4454