Learn More
Sino Biological Recombinant Human CD32B/Fcgr2b Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10259H08C100
Encompass_Preferred
A DNA sequence encoding (Ala46-Pro217) of human CD32b (NP_001002274.1) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4456