Learn More
Sino Biological Recombinant Human CD32B/Fcgr2b Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10259H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Ala 46-Pro 217) of mature human CD32b (NP_001002274.1) protein was expressed with a N-terminal signal peptide and a C-terminal polyhistidine tag.
Catalog No. 50-161-4458