Learn More
Sino Biological Anti-CD32B/Fcgr2b Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10259MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD32b / FCGR2B / Fc gamma RIIB (rh CD32b / FCGR2B / Fc gamma RIIB; Catalog#10259-H08H; NP_001002274.1; Ala46-Pro217). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3777