Learn More
Sino Biological Anti-CD32B/Fcgr2b Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10259MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD32b extracellular domain (rhCD32b; Catalog#10259-H08H; Ala 46-Pro 217; NP_001002274.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-3780